missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL36 Beta/IL1F8 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18043-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
IL-36B is is a member of the interleukin 1 cytokine family whose gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. IL-36B is thought to activate the NF-kappaB pathway through IL-1 receptor family members IL-1RL2 and IL-1RAcP. Like the related proteins IL-36A and IL-36G, IL-36B requires post-translational processing for full agonist activity, but the cleavage mechanism is currently unknown. The IL-36 cytokines have been suggested to amplify Th1 responses by enhancing proliferation and Th1 polarization of naive CD4+ T cells.Specifications
IL36 Beta/IL1F8 | |
Polyclonal | |
Unconjugated | |
IL36B | |
FIL1, FIL1 (ETA), FIL1 eta, FIL1H, IL 1 eta, IL 1F8, IL 1H2, IL1 ETA, IL1F8, IL1H2, Interleukin 1 eta, Interleukin 1 family member 8, Interleukin 1 homolog 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27177 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9NZH7 | |
IL36B | |
IL36 Beta/IL1F8 Fusion Protein Ag12675 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |