missing translation for 'onlineSavingsMsg'
Learn More

IL3RA Recombinant Human Protein, hIgG1-Fc.His Tag

IL3RA Recombinant Human Protein, hIgG1-Fc.His Tag

Brand:  Invitrogen 10518H03H25

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15276865

  • 3575.00 SEK / 25µg


Estimated Shipment date
from Supplier 02-04-2019
Add to basket

Description

Description

Interleukin 3 receptor, alpha (low affinity) (IL3RA), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the human IL3RA (NP_002174.1) extracellular domain (Met 1-Arg 305) fused to the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.

Activity: Measured by its binding ability in a functional ELISA
. Immobilized human IL3 at 20μg/ml (100μl/well) can bind human IL3RA with a linear range of 0.064-1.6μg/ml.

Formulation: Lyophilized in 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Protein assays and analysis
Lyophilized
IL3RA Recombinant Human Protein
5 x 5μg
For research use only. Not for use in diagnostic procedures
Cytokine receptors, interleukins
IL3RA Recombinant Human Protein
IL3RA
Cytokines and receptors
IL3RA Recombinant Human Protein
His Tag, IgG1 Fc Tag, hIgG1-Fc Tag
Lyophilized
61kDa
≥85% by SDS-PAGE
–20°C
5 Pack(s)
IL3RA, RP11-261P4.2, CD123, IL3R, IL3RAY, IL3RX, IL3RY, MGC34174, hIL-3Ra
5 Vials of lyophilized protein
Assay standard
Mammalian
Human
Videos