missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
IL6R Antibody (RM0114-14F33), Alexa Fluor™ 350, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-22492AF350
This item is not returnable.
View return policy
Description
IL6R Monoclonal specifically detects IL6R in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| IL6R | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| IL6R | |
| Mouse recombinant protein of the extracellular domain of IL-6 R | |
| 0.1 mL | |
| 3570 | |
| Mouse | |
| IgG2 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| RM0114-14F33 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen | |
| CD126, CD126 antigen, gp80, IL-6 receptor subunit alpha, IL-6R 1, IL-6R subunit alpha, IL-6R-1, IL6RA, IL-6RA, IL-6R-alpha, interleukin 6 receptor, interleukin 6 receptor alpha subunit, interleukin-6 receptor subunit alpha, Membrane glycoprotein 80, MGC104991 | |
| Rat | |
| Protein A or G purified | |
| Primary | |
| This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of the extracellular domain of IL-6 R. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction