missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ILK (Human) Recombinant Protein
Human ILK full-length ORF ( NP_001014794.1, 1 a.a. - 452 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00003611-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MDDIFTQCREGNAVAVRLWLDNTENDLNQGDDHGFSPLHWACREGRSAVVEMLIMRGARINVMNRGDDTPLHLAASHGHRDIVQKLLQYKADINAVNEHGNVPLHYACFWGQDQVAEDLVANGALVSICNKYGEMPVDKAKAPLRELLRERAEKMGQNLNRIPYKDTFWKGTTRTRPRNGTLNKHSGIDFKQLNFLTKLNENHSGELWKGRWQGNDIVVKVLKVRDWSTRKSRDFNEECPRLRIFSHPNVLPVLGACQSPPAPHPTLITHWMPYGSLYNVLHEGTNFVVDQSQAVKFALDMARGMAFLHTLEPLIPRHALNSRSVMIDEDMTARISMADVKFSFQCPGRMYAPAWVAPEALQKKPEDTNRRSADMWSFAVLLWELVTREVPFADLSNMEIGMKVALEGLRPTIPPGISPHVCKLMKICMNEDPAKRPKFDMIVPILEKMQDK
Specifications
NP_001014794.1 | |
integrin-linked kinase | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ILK | |
GST |
3611 | |
Wheat germ expression system | |
10 μg | |
DKFZp686F1765, P59 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |