Läs mer
Imaradenant, MedChemExpress
Produktkod. 16476407
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16476407 5 mg 5mg
16403678 10 mg -
16400067 50 mg -
16404756 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16476407 Leverantör Medchem Express Leverantörsnummer HY101980
MedChemExpress AZD4635 (HTL1071) is a potent, selective and orally active adenosine A2A receptor (A2AR) antagonist. AZD4635 binds to human A2AR with a Ki of 1.7 nM and shows >30-fold selectivity over other adenosine receptors.
AZD4635 (HTL1071) is a potent, selective and orally active adenosine A2A receptor (A2AR) antagonist. AZD4635 binds to human A2AR with a Ki of 1.7 nM and shows >30-fold selectivity over other adenosine receptors.
Specifikationer
Kemiskt namn eller material Imaradenant
CAS 1321514-06-0
Färg Light Yellow
Anteckningar om renhetsgrad Research
Molekylformel C15H11ClFN5
Kvantitet 5 mg
Synonym HTL1071 AZD4635
Löslighetsinformation DMSO : ≥ 83.3 mg/mL (263.83 mM)
LEDER NC1=NC(C2=CC=C(F)C=C2)=C(C3=CC(Cl)=NC(C)=C3)N=N1
Molekylvikt (g/mol) 315.73
Formel vikt 315.73
Procent renhet 98.85%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-programmed cell death