Läs mer
Imazamethabenz-methyl, TRC
Produktkod. 30137033
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137033 25 mg
17980585 5 mg
17961695 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137033 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI26863025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 81405-85-8
Molekylformel C16 H20 N2 O3
Molekylvikt (g/mol) 288.34
Synonym Benzoic acid, 2-[4,5-dihydro-4-methyl-4-(1-methylethyl)-5-oxo-1H-imidazol-2-yl]-4(or 5)-methyl-, methyl ester (9CI, ACI), AC 222293, Assert, Assert 300, Dagger, Dagger G, Imazamethabenz methyl ester, Imazamethabenz-methyl
IUPAC-namn methyl 4-methyl-2-(4-methyl-5-oxo-4-propan-2-yl-1H-imidazol-2-yl)benzoate;methyl 5-methyl-2-(4-methyl-5-oxo-4-propan-2-yl-1H-imidazol-2-yl)benzoate
LEDER COC(=O)c1ccc(C)cc1C2=NC(C)(C(C)C)C(=O)N2.COC(=O)c3cc(C)ccc3C4=NC(C)(C(C)C)C(=O)N4
Specifikationer
Kemiskt namn eller material Imazamethabenz-methyl
Kvantitet 25 mg
InChI formel InChI=1S/2C16H20N2O3/c1-9(2)16(4)15(20)17-13(18-16)12-8-10(3)6-7-11(12)14(19)21-5;1-9(2)16(4)15(20)17-13(18-16)11-7-6-10(3)8-12(11)14(19)21-5/h2*6-9H,1-5H3,(H,17,18,20)
Formel vikt 576.2948