Läs mer
Imazamox Methyl Ester, TRC
Produktkod. 30406841
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30406841 1 g
1 options
Produktkod. 30406841 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI2686001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Imazamox-methyl
CAS 114526-46-4
Molekylformel C16 H21 N3 O4
Kvantitet 1 g
Synonym 3-Pyridinecarboxylic acid, 2-[4,5-dihydro-4-methyl-4-(1-methylethyl)-5-oxo-1H-imidazol-2-yl]-5-(methoxymethyl)-, methyl ester, Methyl 2-[4,5-dihydro-4-methyl-4-(1-methylethyl)-5-oxo-1H-imidazol-2-yl]-5-(methoxymethyl)-3-pyridinecarboxylate
InChI formel InChI=1S/C16H21N3O4/c1-9(2)16(3)15(21)18-13(19-16)12-11(14(20)23-5)6-10(7-17-12)8-22-4/h6-7,9H,8H2,1-5H3,(H,18,19,21)
LEDER COCc1cnc(C2=NC(C)(C(C)C)C(=O)N2)c(c1)C(=O)OC
IUPAC-namn methyl 5-(methoxymethyl)-2-(4-methyl-5-oxo-4-propan-2-yl-1H-imidazol-2-yl)pyridine-3-carboxylate
Molekylvikt (g/mol) 319.36
Formel vikt 319.1532