Läs mer
Imazethapyr, TRC
Produktkod. 30179995
Change view
Kvantitet:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179995 25 g
30179869 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30179995 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI26862525G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 81335-77-5
Molekylformel C15 H19 N3 O3
Molekylvikt (g/mol) 289.33
Synonym 2-[4,5-Dihydro-4-methyl-4-(1-methylethyl)-5-oxo-1H-imidazol-2-yl]-5-ethyl-3-pyridinecarboxylic acid, (±)-5-Ethyl-2-(4-isopropyl-4-methyl-5-oxo-4,5-dihydro-1H-imidazol-2-yl)nicotinic acid, (±)-Imazethapyr, 5-Ethyl-2-(4-isopropyl-4-methyl-5-oxo-2-imidazolin-2-yl) nicotinic acid, AC 263499, CL 263499, Fabian, Imazethapyr, Pivot, Pursuit, Pursuit 10
IUPAC-namn 5-ethyl-2-(4-methyl-5-oxo-4-propan-2-yl-1H-imidazol-2-yl)pyridine-3-carboxylic acid
LEDER CCc1cnc(C2=NC(C)(C(C)C)C(=O)N2)c(c1)C(=O)O
Specifikationer
Kemiskt namn eller material Imazethapyr
Kvantitet 25 g
InChI formel InChI=1S/C15H19N3O3/c1-5-9-6-10(13(19)20)11(16-7-9)12-17-14(21)15(4,18-12)8(2)3/h6-8H,5H2,1-4H3,(H,19,20)(H,17,18,21)
Formel vikt 289.1426