Läs mer
Imeglimin hydrochloride, MedChemExpress
Produktkod. 16486158
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16486158 10 mg -
16468757 5 mg -
16462297 50 mg 50mg
16456546 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16486158 Leverantör Medchem Express Leverantörsnummer HY14771A
MedChemExpress Imeglimin hydrochloride (EMD 387008) is an oral glucose-lowering agent. Imeglimin also reduces reactive oxygen species (ROS) production, increases mitochondrial DNA and improves mitochondrial function.
Imeglimin hydrochloride (EMD 387008) is an oral glucose-lowering agent. Imeglimin also reduces reactive oxygen species (ROS) production, increases mitochondrial DNA and improves mitochondrial function.
Specifikationer
Kemiskt namn eller material Imeglimin hydrochloride
Fysisk form Solid
CAS 775351-61-6
Färg Off-White
Kvantitet 10 mg
Anteckningar om renhetsgrad Research
Molekylformel C6H14ClN5
För användning med (applikation) Metabolism-sugar/lipid metabolism
Synonym EMD 387008 hydrochloride
Löslighetsinformation H2O : ≥ 50 mg/mL (260.88 mM) ∣DMSO : 25 mg/mL (130.44 mM; Need ultrasonic)
LEDER [H]Cl.NC1=N[C@@H](C)N=C(N(C)C)N1
Molekylvikt (g/mol) 191.66
Formel vikt 191.66
Procent renhet 98.0%