Läs mer
Imidacloprid-olefin, TRC
Produktkod. 30132437
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132437 100 mg
17951515 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30132437 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI274995100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 115086-54-9
Molekylformel C9 H10 Cl N5 O2
Molekylvikt (g/mol) 254.65
Synonym 1-[(6-Chloro-3-pyridinyl)methyl]-N-nitro-1H-imidazol-2-amine, Imidacloprid-olefin, NTN 33893-olefin, NTN 35884
IUPAC-namn (NE)-N-[3-[(6-chloropyridin-3-yl)methyl]-1H-imidazol-2-ylidene]nitramide
LEDER [O-][N+](=O)\N=C\1/NC=CN1Cc2ccc(Cl)nc2
Specifikationer
Kemiskt namn eller material Imidacloprid-olefin
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C9H8ClN5O2/c10-8-2-1-7(5-12-8)6-14-4-3-11-9(14)13-15(16)17/h1-5H,6H2,(H,11,13)
Formel vikt 253.0367
Procent renhet >95%