Läs mer
Imidacloprid-urea, TRC
Produktkod. 30135827
Change view
Kvantitet:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135827 500 mg
30133038 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30135827 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI274985500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 120868-66-8
Molekylformel C9 H10 Cl N3 O
Molekylvikt (g/mol) 211.65
Synonym 1-[(6-Chloro-3-pyridinyl)methyl]-2-imidazolidinone, Imidacloprid-urea
IUPAC-namn 1-[(6-chloropyridin-3-yl)methyl]imidazolidin-2-one
LEDER Clc1ccc(CN2CCNC2=O)cn1
Specifikationer
Kemiskt namn eller material Imidacloprid-urea
Anteckningar om renhetsgrad HPLC
Kvantitet 500 mg
InChI formel InChI=1S/C9H10ClN3O/c10-8-2-1-7(5-12-8)6-13-4-3-11-9(13)14/h1-2,5H,3-4,6H2,(H,11,14)
Formel vikt 211.0512
Procent renhet >95%