Learn More
Imidazol-1-yl-acetic Acid, TRC
Product Code. 30396230
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity Unit Size
30396230 1 g 1g
1 options
Product Code. 30396230 Supplier Toronto Research Chemicals Supplier No. TRCI3856001G
 Restricted Item
Beräknad leverans: 09-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Imidazol-1-yl-acetic Acid
CAS 22884-10-2
Molecular Formula C5 H6 N2 O2
Quantity 1 g
Synonym 2-(1H-Imidazol-1-yl)acetic Acid, Imidazole-1-acetic acid (6CI,7CI,8CI), (1H-Imidazol-1-yl)acetic acid, 2-(1H-Imidazol-1-yl)acetic acid, Imidazol-1-ylacetic acid
InChI Formula InChI=1S/C5H6N2O2/c8-5(9)3-7-2-1-6-4-7/h1-2,4H,3H2,(H,8,9)
SMILES OC(=O)Cn1ccnc1
IUPAC Name 2-imidazol-1-ylacetic acid
Molecular Weight (g/mol) 126.11
Formula Weight 126.0429