missing translation for 'onlineSavingsMsg'
Learn More

Merck Millipore Imidazole

Buffer substance ACS

527.00 SEK - 4124.00 SEK

Specifications

Assay ≥99% (perchloric acid titration)
CAS 288-32-4
Form Solid
Formulation C3H4N2
Molecular Weight 68.08
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10317190
View Documents
Merck Millipore
1047160050
50 g 527.00 SEK
50g
Qty Price Break Available View alternative products
Save Up to 
»
Estimated Shipment: 05-08-2021
Please to purchase this item. Need a web account? Register with us today!
10580781
View Documents
Merck Millipore
1047160250
250 g
1401.00 SEK
250g
View alternative products
Save Up to 
»
Estimated Shipment: 11-08-2021
Please to purchase this item. Need a web account? Register with us today!
10290741
View Documents
Merck Millipore
1047161000
1 kg 4124.00 SEK
1kg
Qty Price Break Available View alternative products
Save Up to 
»
Estimated Shipment: 11-08-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Synonym: Glyoxaline, Iminazole, Imidazole
  • Grade: ACS reagent
  • Assay (perchloric acid titration): ≥ 99
  • Identity (IR-spectrum): conforms
  • pH-value (5 %, in water): 9.5 - 11.0
  • Melting point: 90 - 92
  • UV/VIS Spectrum: conforms
  • Fe (Iron): ≤ 0.001 %
  • Sulfated ash: ≤ 0.1
  • Water (according to Karl Fischer): ≤ 0.2
  • Corresponds to ACS
Specifications

Specifications

≥99% (perchloric acid titration)
Solid
68.08
Store at 15°C to 25°C
288-32-4
C3H4N2
10.5 (20°C, 67 g/L in H2O)
SDS
Product Certifications