Learn More
Imidazole, >98.5% (Dry Basis), Ultrapure, Thermo Scientific Chemicals
Catalog Number p-7082797
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
Plastic bottle
288-32-4
C3H4N2
68.08
MFCD00005183
RAXXELZNTBOGNW-UHFFFAOYSA-N
imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole
795
CHEBI:16069
1 H-imidazole
N1C=CN=C1
288-32-4
C3H4N2
68.08
MFCD00005183
RAXXELZNTBOGNW-UHFFFAOYSA-N
imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole
795
CHEBI:16069
1 H-imidazole
N1C=CN=C1
Melting Point 88°C to 92°C
Density 1.03
Boiling Point 256°C
Flash Point 145°C
Packaging Plastic bottle
Quantity 100 g
UN Number UN3263
Beilstein 103853
Specific Gravity 1.03
Formula Weight 68.08
Percent Purity ≥98.5%
Grade Ultrapure
Physical Form Flakes or Powder
Assay Dry basis
Chemical Name or Material Imidazole