missing translation for 'onlineSavingsMsg'
Learn More

Imide-extended bismaleimide, BMI-1500, Thermo Scientific Chemicals

Catalog Number 15452787
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
1290041-56-3
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
1290041-56-3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Amber
Physical Form Viscous Liquid
Quantity 100 g
Chemical Name or Material Imide-extended bismaleimide, BMI-1500

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'