missing translation for 'onlineSavingsMsg'
Learn More

Imide-extended bismaleimide, powder, BMI-3000, Thermo Scientific™

Catalog Number p-7080175
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
921213-77-6
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
921213-77-6

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Yellow
Physical Form Powder
Quantity 25 g
Chemical Name or Material Imide-extended bismaleimide, BMI-3000
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'