Läs mer
Imiquimod USP Related Compound A, Mikromol™
Produktkod. 30460480
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30460480 4 x 25 mg
30050624 100 mg
30051099 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30460480 Leverantör Mikromol Leverantörsnummer MM1158.0200254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Imiquimod USP Related Compound A, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Desamino Imiquimod
CAS 99010-24-9
Analyt- eller komponentnamn Desamino Imiquimod
Frakt skick Room Temperature
Molekylformel C14 H15 N3
Certifikat och efterlevnad ISO 9001
Synonym 1-Isobutyl-1H-imidazo[4,5-c]quinoline (Desaminoimiquimod), 1H-Imidazo[4,5-c]quinoline, 1-(2-methylpropyl)-, 1-(2-Methylpropyl)-1H-imidazo[4,5-c]quinoline, 1-Isobutyl-1H-imidazo-[4,5-c]-quinoline, 1-Isobutyl-1H-imidazo[4,5-c]quinoline, Imiquimod Related Compound A (USP), Imiquimod Related Compound A
InChI formel InChI=1S/C14H15N3/c1-10(2)8-17-9-16-13-7-15-12-6-4-3-5-11(12)14(13)17/h3-7,9-10H,8H2,1-2H3
Lösningstyp Neat
LEDER CC(C)Cn1cnc2cnc3ccccc3c12
IUPAC-namn 1-(2-methylpropyl)imidazo[4,5-c]quinoline
Molekylvikt (g/mol) 225.29
Kvantitet 4 x 25 mg
Formel vikt 225.1266 g/mol