missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Imject™ Freund's Adjuvants

Stimulate a nonspecific immune response using complete (CFA) or incomplete (IFA) formulations of the popular Freund's adjuvant; offered in 10mL ampules.

806.00 SEK - 884.00 SEK

Specifications

Quantity 5 x 10mL
Sufficient For Mixture with 50mL of immunogen
View More Specs

Products 2
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
11824071 View Documents Thermo Scientific™
77140
Imject Freund's Complete Adjuvant (FCA) 884.00 SEK
Pack of 5 View alternative products
Save Up to 
»
EU Stock 12
Add to basket
 
11834071 View Documents Thermo Scientific™
77145
Imject Freund's Incomplete Adjuvant (FIA) 806.00 SEK
Pack of 5 View alternative products
Save Up to 
»
EU Stock 19
Add to basket
 
Description

Description

The water-and-oil emulsions (with and without inactivated Mycobacterium) greatly enhance immune responses to immunogens when mixed and injected with the prepared antigen. Typically, FCA is used for initial injections and FIA is used for booster injections. The ampules comprise conveniently sized aliquots that provide long shelf life.

Highlights:

  • Water-in-oil emulsion with or without killed Mycobacterium
  • Enhance immune response and antibody production for injected immunogens
  • Useful for immunization of mice, rabbits and other mammalian hosts
  • Requires emulsification with prepared immunogens

Freund's Complete Adjuvant (FCA), Part No. 77140:

  • Water-in-oil emulsion containing killed Mycobacterium
  • Greatly enhances immune response to immunogen
  • Typically used for initial injections
  • Disadvantages: difficult to mix with immunogen and can cause tissue necrosis at injection site

Freund's Incomplete Adjuvant (FIA), Part No. 77145:

  • Water-in-oil emulsion without Mycobacterium
  • Enhances immune response in booster injections
  • Typically used for subsequent boosts after initial injection with FCA
  • Disadvantage: difficult to mix with immunogen
Specifications

Specifications

5 x 10mL
Mixture with 50mL of immunogen