Learn More
Fisherbrand™ Immersion Basket with Handle
Faciliates complete immersion within S-line Ultrasonic Cleaner baths. Fisherbrand™ Immersion Baskets with Handle are manufactured from stainless steel.
Brand:  Fisherbrand™
Additional Details : Weight : 1.34100kg
 View more versions of this product
Product Code. 11786188
Quantity Price
1 1748.00 SEK / Each
10 1612.70 SEK / Each
In stock, ready to ship within 7 days: 1
Add to basket
Description
Description
  • Mesh bottom
  • One handle on each end
  • Functional and modern design
  • Available for a variety of capacities
Specifications
Specifications
9.5 L Elmasonic S 100 (H) Ultrasonic Baths
76.2 mm
Stainless Steel
Basket
254 mm
200.66 mm