missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROBU VitraPOR™ Immersion Filter
Immersion Filter for reverse filtration.
Brand: ROBU VitraPOR™ 24350
Additional Details : Weight : 0.13200kg
Description
- Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Borosilicate Glass 3.3 | |
35 mm | |
0 |
Non-sterile | |
210 mm |