missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IMP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | IMP3 |
---|---|
Concentration | 0.16 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16850634
|
Proteintech
12750-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16840634
|
Proteintech
12750-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes the human homolog of the yeast Imp3 protein. The protein localizes to the nucleoli and interacts with the U3 snoRNP complex. The protein contains an S4 domain.Specifications
IMP3 | |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q921Y2, Q9NV31 | |
102462, 315697, 55272 | |
IMP3 Fusion Protein Ag3477 | |
Primary | |
-20°C | |
IMP3 |
0.16 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
BRMS2, C15orf12, IMP3, MRPS4, U3 snoRNP protein IMP3 | |
Imp3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |