missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ IMP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531769
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Recommended positive controls: Raji. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (82%), Rhesus Monkey (100%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
IMP4 forms a ternary complex with IMP3.
Specifikationer
| IMP4 | |
| Polyclonal | |
| Unconjugated | |
| Imp4 | |
| AA409888; AV031295; Brix domain-containing protein 4; BXDC4; D1Wsu40e; IMP4; IMP4 homolog, U3 small nucleolar ribonucleoprotein; IMP4, U3 small nucleolar ribonucleoprotein; IMP4, U3 small nucleolar ribonucleoprotein, homolog; IMP4, U3 small nucleolar ribonucleoprotein, homolog (yeast); LRRGT00017; RGD1308463; U3 small nucleolar ribonucleoprotein protein IMP4; U3 snoRNP protein 4 homolog; U3 snoRNP protein IMP4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 92856 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q96G21 | |
| Imp4 | |
| Recombinant protein encompassing a sequence within the center region of human IMP4. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering