Learn More
Unity Lab Services™ Incubator Requalification Service
Product Code. 30178680
Change view
Click to view available options
For Use With (Equipment):
Incubator
Unit Size:
1 unit
Product Code. For Use With (Equipment) Unit Size
30178680 Incubator 1 unit
1 options
Product Code. 30178680 Supplier Unity Lab Services™ Supplier No. RQINC
 Restricted Item
Beräknad leverans: 21-12-2026
Requalification Services for Incubator (documentation included)
Requalification (RQ): Documented verification that equipment (as installed) is operating as intended according to manufacturers specifications.
  • Verifies important equipment functions and helps ensure that the equipment operates as expected by the manufacturer and conforms to standards and requirements.
  • Key tests include door ajar, power failure, and temperature alerts.
  • Please note that installation and operational qualification, ideally along with preventive maintenance and calibration, should be completed on lab equipment before requalification services can be performed.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Incubator
Product Type Requalification Service