Läs mer
Incyclinide, MedChemExpress
Produktkod. 16461947
Encompass_Preferred
Change view
Kvantitet:
1 mg
5 mg
10 mg
50 mg
Förpackningsstorlek:
1mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16461947 50 mg -
16461369 1 mg 1mg
16438467 5 mg -
16435798 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16461947 Leverantör Medchem Express Leverantörsnummer HY13648
MedChemExpress Incyclinide (CMT-3, COL-3) is a matrix metalloproteinase (MMP) inhibitor, thereby inducing extracellular matrix degradation, and inhibiting angiogenesis, tumor growth and invasion, and metastasis.
Incyclinide (CMT-3, COL-3) is a matrix metalloproteinase (MMP) inhibitor, thereby inducing extracellular matrix degradation, and inhibiting angiogenesis, tumor growth and invasion, and metastasis.
Specifikationer
Kemiskt namn eller material Incyclinide
CAS 15866-90-7
Färg Off-White
Anteckningar om renhetsgrad Research
Molekylformel C19H17NO7
Kvantitet 50 mg
Synonym CMT-3 COL-3
Löslighetsinformation DMSO : ≥ 100 mg/mL (269.29 mM)
LEDER O=C(C(C1=O)=C(O)C[C@]2([H])C[C@]3([H])CC4=C(C(C3=C(O)[C@@]21O)=O)C(O)=CC=C4)N
Molekylvikt (g/mol) 371.34
Formel vikt 371.34
Procent renhet 97.06%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease