Learn More
Indapamide
Product Code. 15446009
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15446009 5 g 5g
15436009 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15446009 Supplier Thermo Scientific Alfa Aesar Supplier No. J63846.06
 Restricted Item
I lager
Add to basket
CAS: 26807-65-8 | C16H16ClN3O3S | 365.83 g/mol
Used as an antihypertensive. Diuretic
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Used as an antihypertensive
  • Diuretic
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 26807-65-8
Molecular Formula C16H16ClN3O3S
Molecular Weight (g/mol) 365.83
MDL Number MFCD00079375
InChI Key NDDAHWYSQHTHNT-UHFFFAOYNA-N
Synonym indapamide, noranat, veroxil, tertensif, arifon, fludex, lozol, indaflex, bajaten, indamol
PubChem CID 3702
ChEBI CHEBI:5893
IUPAC Name 4-chloro-N-(2-methyl-2,3-dihydro-1H-indol-1-yl)-3-sulfamoylbenzamide
SMILES CC1CC2=CC=CC=C2N1NC(=O)C1=CC=C(Cl)C(=C1)S(N)(=O)=O
Specifications
Melting Point 160°C to 162°C
Color White
Quantity 5 g
Merck Index 14,4935
Solubility Information Soluble in ethanol; Insoluble in water
Formula Weight 365.83
Physical Form Powder
Chemical Name or Material Indapamide