Läs mer
Tocris Bioscience™ Indatraline hydrochloride
GREENER_CHOICE
Produktkod. 18095652
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18095652 10 mg
18005352 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18095652 Leverantör Tocris Bioscience™ Leverantörsnummer 1588/10
Potent 5-HT uptake inhibitor; also inhibits dopamine and noradrenalin uptake
Indatraline hydrochloride is a potent monoamine uptake inhibitor. Inhibits transporters for 5-HT (SERT), dopamine (DAT) and noradrenaline (NET) (Ki values are 0.42, 1.7 and 5.8 nM respectively). Centrally active following systemic administration in vivo.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 96850-13-4
Molekylformel C16H16Cl3N
Specifikationer
Kemiskt namn eller material Indatraline hydrochloride
Kvantitet 10 mg
Formel vikt 328.67
Renhet >99%
Molekylvikt (g/mol) 328.661