Läs mer
Indazole-3-carboxylic Acid Methyl Ester, TRC
Produktkod. 30403895
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30403895 5 g
1 options
Produktkod. 30403895 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI5052505G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Indazole-3-carboxylic Acid Methyl Ester
CAS 43120-28-1
Molekylformel C9H8N2O2
Kvantitet 5 g
Synonym 3-(Methoxycarbonyl)-1H-indazole, Methyl 1H-indazole-3-carboxylate, Methyl 3-indazolecarboxylate
InChI formel InChI=1S/C9H8N2O2/c1-13-9(12)8-6-4-2-3-5-7(6)10-11-8/h2-5H,1H3,(H,10,11)
LEDER COC(=O)c1n[nH]c2ccccc12
Molekylvikt (g/mol) 176.17
Formel vikt 176.0586