Learn More
Thermo Scientific Chemicals India Ink soln., 0.2% in PBS buffer
India ink is a simple black ink. Previously it has been used for writing and printing and is now commonly used for drawing in comic books and comic strips.
1075.69 SEK - 1810.57 SEK
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15423679
|
Thermo Scientific Chemicals
J61007.AP |
500mL |
1075.69 SEK
500mL |
In Stock
Log in to see stock. |
|||||
15433679
|
Thermo Scientific Chemicals
J61007.K2 |
1L |
1810.57 SEK
1 L |
In Stock
Log in to see stock. |
|||||
Description
India ink is a simple black ink. Previously it has been used for writing and printing and is now commonly used for drawing in comic books and comic strips. India ink is also used in medical research to visualize blood vessels when viewed under a microscope. It is used in a diluted form as an ultra-fine polishing medium for making precise optical surfaces on metals. In ophthalmology, it is used to some extent in corneal tattooing.