Läs mer
Indinavir Sulfate, TRC
Produktkod. 17971035
Change view
Kvantitet:
5 mg
25 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17971035 1 g
17913825 5 mg
17921725 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17971035 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI5250001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 157810-81-6
Molekylformel C36 H47 N5 O4 . H2 O4 S
Molekylvikt (g/mol) 711.87
Synonym Indinavir sulfate, (2S)-1-[(2S,4R)-4-Benzyl-2-hydroxy-5-[[(1S,2R)-2-hydroxy-2,3-dihydro-1H-inden-1-yl]amino]-5-oxopentyl]-N-(1,1- dimethylethyl)-4-(pyridin-3-ylmethyl)piperazine-2-carboxamide sulfate ethanolate
IUPAC-namn (2S)-1-[(2S,4R)-4-benzyl-2-hydroxy-5-[[(1S,2R)-2-hydroxy-2,3-dihydro-1H-inden-1-yl]amino]-5-oxopentyl]-N-tert-butyl-4-(pyridin-3-ylmethyl)piperazine-2-carboxamide;sulfuric acid
LEDER CC(C)(C)NC(=O)[C@@H]1CN(Cc2cccnc2)CCN1C[C@@H](O)C[C@@H](Cc3ccccc3)C(=O)N[C@@H]4[C@H](O)Cc5ccccc45.OS(=O)(=O)O
Specifikationer
Kemiskt namn eller material Indinavir sulfate
Kvantitet 1 g
InChI formel InChI=1S/C36H47N5O4.H2O4S/c1-36(2,3)39-35(45)31-24-40(22-26-12-9-15-37-21-26)16-17-41(31)23-29(42)19-28(18-25-10-5-4-6-11-25)34(44)38-33-30-14-8-7-13-27(30)20-32(33)43;1-5(2,3)4/h4-15,21,28-29,31-33,42-43H,16-20,22-24H2,1-3H3,(H,38,44)(H,39,45);(H2,1,2,3,4)/t28-,29+,31+,32-,33+;/m1./s1
Formel vikt 711.3302