Läs mer
Indirubin, MedChemExpress
Produktkod. 16478986
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
50 mg
100 mg
250 mg
Förpackningsstorlek:
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16478986 100 mg -
16473836 25 mg -
16471308 5 mg 5mg
16469895 250 mg -
16468868 10 mg -
16464757 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Produktkod. 16478986 Leverantör Medchem Express Leverantörsnummer HYN0117
MedChemExpress Indirubin (Couroupitine B) is a bis-indole alkaloid and has emarkable anticancer activity against chronic myelocytic leukemia.
Indirubin (Couroupitine B) is a bis-indole alkaloid and has emarkable anticancer activity against chronic myelocytic leukemia.
Specifikationer
Kemiskt namn eller material Indirubin
CAS 479-41-4
Färg Reddish Brown
Anteckningar om renhetsgrad Research
Molekylformel C16H10N2O2
Kvantitet 100 mg
Synonym Couroupitine B Indigo red Indigopurpurin
Löslighetsinformation DMSO : 5 mg/mL (19.07 mM; ultrasonic and warming and heat to 60°C)
LEDER O=C1NC2=C(C=CC=C2)/C1=C3NC4=C(C=CC=C4)C\3=O
Molekylvikt (g/mol) 262.26
Formel vikt 262.26
Procent renhet 96.0%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease