missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) trifluoromethanesulfonate, 98%, Thermo Scientific™

1016.46 SEK - 3442.92 SEK

Chemical Identifiers

CAS 128008-30-0
Molecular Formula C3F9InO9S3
Molecular Weight (g/mol) 562.03
MDL Number MFCD00144478
InChI Key UCYRAEIHXSVXPV-UHFFFAOYSA-K
Synonym indium iii trifluoromethanesulfonate, indium triflate, indium trifluoromethanesulphonate, indium 3+ tritriflate, indium 3+ ; trifluoromethanesulfonate, indium iii triflate, indium iii trifluoromethanesulphonate, acmc-209bda, in tfa 3, ksc491q8t
PubChem CID 2734680
IUPAC Name indium(3+);trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[In+3]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10643773
SDS View Documents
Thermo Scientific Acros
368540050
5g Glass bottle
1016.46 SEK
5g
View alternative products
Save Up to 
»
In stock, ready to ship: 11
Please to purchase this item. Need a web account? Register with us today!
10521902
SDS View Documents
Thermo Scientific Acros
368540250
25g Glass bottle
3442.92 SEK
25g
View alternative products
Save Up to 
»
Estimated Shipment: 11-11-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

128008-30-0
562.03
UCYRAEIHXSVXPV-UHFFFAOYSA-K
2734680
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[In+3]
C3F9InO9S3
MFCD00144478
indium iii trifluoromethanesulfonate, indium triflate, indium trifluoromethanesulphonate, indium 3+ tritriflate, indium 3+ ; trifluoromethanesulfonate, indium iii triflate, indium iii trifluoromethanesulphonate, acmc-209bda, in tfa 3, ksc491q8t
indium(3+);trifluoromethanesulfonate

Specifications

Indium(III) trifluoromethanesulfonate
Authentic
White
128008-30-0
C3F9InO9S3
indium iii trifluoromethanesulfonate, indium triflate, indium trifluoromethanesulphonate, indium 3+ tritriflate, indium 3+ ; trifluoromethanesulfonate, indium iii triflate, indium iii trifluoromethanesulphonate, acmc-209bda, in tfa 3, ksc491q8t
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[In+3]
562.03
562.03
98%
97.5% min. (ex Fluor)
Glass bottle
5g
98%
MFCD00144478
UCYRAEIHXSVXPV-UHFFFAOYSA-K
indium(3+);trifluoromethanesulfonate
2734680
Powder and Granules
Safety and Handling

Safety and Handling

Product Identifier
  • Indium(III) trifluoromethanesulfonate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Product Certifications

RUO – Research Use Only