missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indole-2-carboxaldehyde, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
Indole-2-carboxaldehyde is a useful starter in indole and natural product chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Indole-2-carboxaldehyde is a useful starter in indole and natural product chemistry.
Solubility
Soluble in Methanol.
Notes
Air sensitive. Strong oxidizers and strong reducing agents are incompatible. Store at room temperature.
Indole-2-carboxaldehyde is a useful starter in indole and natural product chemistry.
Solubility
Soluble in Methanol.
Notes
Air sensitive. Strong oxidizers and strong reducing agents are incompatible. Store at room temperature.
Specifications
Melting Point | 138°C to 142°C |
Quantity | 250 mg |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in Methanol. |
Formula Weight | 145.16 |
Percent Purity | 97% |
Chemical Name or Material | Indole-2-carboxaldehyde |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'