Läs mer
Indole-3-acetyl-L-glutamic Acid, TRC
Produktkod. 30010407
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010407 10 mg
30007134 1 mg
30006089 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010407 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI57737510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 57105-48-3
Molekylformel C15H16N2O5
Molekylvikt (g/mol) 304.3
Synonym N-[2-(1H-Indol-3-yl)acetyl]-L-glutamic Acid, N-(Indol-3-ylacetyl)-glutamic Acid, Indole-3-acetyl Glutamate, Indole-3-acetylglutamic Acid;
LEDER C1=CC=C2C(=C1)C(=CN2)CC(=N[C@@H](CCC(=O)O)C(=O)O)O
Specifikationer
Kemiskt namn eller material Indole-3-acetyl-L-glutamic Acid
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C15H16N2O5/c18-13(17-12(15(21)22)5-6-14(19)20)7-9-8-16-11-4-2-1-3-10(9)11/h1-4,8,12,16H,5-7H2,(H,17,18)(H,19,20)(H,21,22)/t12-/m0/s1
Kvantitet 10 mg
Procent renhet >95%