Läs mer
Indomethacin 1-Menthol ester, TRC
Product Code. 30404097
Change view
Click to view available options
Kvantitet:
1 g
Product Code. Kvantitet
30404097 1 g
1 options
Product Code. 30404097 Supplier Toronto Research Chemicals Supplier No. TRCI1936001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material Indomethacin 1-Menthol ester
CAS 193603-94-0
Molekylformel C29 H34 Cl N O4
Kvantitet 1 g
Synonym Indomethacin Impurity 19
InChI formel InChI=1S/C29H34ClNO4/c1-17(2)23-12-6-18(3)14-27(23)35-28(32)16-24-19(4)31(26-13-11-22(34-5)15-25(24)26)29(33)20-7-9-21(30)10-8-20/h7-11,13,15,17-18,23,27H,6,12,14,16H2,1-5H3/t18-,23+,27-/m1/s1
LEDER COc1ccc2c(c1)c(CC(=O)O[C@@H]3C[C@H](C)CC[C@H]3C(C)C)c(C)n2C(=O)c4ccc(Cl)cc4
IUPAC-namn [(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexyl] 2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetate
Molekylvikt (g/mol) 496.038
Formel vikt 495.218