Läs mer
Indomethacin Ethyl Ester, TRC
Produktkod. 30132513
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132513 25 mg
30009271 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30132513 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI64103025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 16401-99-3
Molekylformel C21 H20 Cl N O4
Molekylvikt (g/mol) 385.84
Synonym 1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indole-3-acetic acid ethyl ester, Indole-3-acetic acid, 1-(p-chlorobenzoyl)-5-methoxy-2-methyl-, ethyl ester (8CI), Ethyl 1-(4-chlorobenzoyl)-5-methoxy-2-methylindole-3-acetate, Ethyl 1-(p-chlorobenzoyl)-2-methyl-5-methoxy-3-indolylacetate, Indomethacin ethyl ester
IUPAC-namn ethyl 2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetate
LEDER CCOC(=O)Cc1c(C)n(C(=O)c2ccc(Cl)cc2)c3ccc(OC)cc13
Specifikationer
Kemiskt namn eller material Indomethacin Ethyl Ester
Kvantitet 25 mg
InChI formel InChI=1S/C21H20ClNO4/c1-4-27-20(24)12-17-13(2)23(19-10-9-16(26-3)11-18(17)19)21(25)14-5-7-15(22)8-6-14/h5-11H,4,12H2,1-3H3
Formel vikt 385.1081