missing translation for 'onlineSavingsMsg'
Learn More

Indomethacin, MP Biomedicals™

Catalog Number p-4679666
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
This item is not returnable. View return policy
53-86-1
C19H16ClNO4
357.79
MFCD00057095
CGIGDMFJXJATDK-UHFFFAOYSA-N
indomethacin, indometacin, indocin, indometacine, indomethacine, indocid, metindol, amuno, indomethazine, imbrilon
3715
CHEBI:49662
2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetic acid
CC1=C(C2=C(N1C(=O)C3=CC=C(C=C3)Cl)C=CC(=C2)OC)CC(=O)O
This item is not returnable. View return policy
53-86-1
C19H16ClNO4
357.79
MFCD00057095
CGIGDMFJXJATDK-UHFFFAOYSA-N
indomethacin, indometacin, indocin, indometacine, indomethacine, indocid, metindol, amuno, indomethazine, imbrilon
3715
CHEBI:49662
2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetic acid
CC1=C(C2=C(N1C(=O)C3=CC=C(C=C3)Cl)C=CC(=C2)OC)CC(=O)O
Molecular biology and Cell biology
Color White
Quantity 1 g
Formula Weight 357.8
Physical Form Solid
Chemical Name or Material Indomethacin
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'