Läs mer
Indoramin Hydrochloride, LoGiCal
Produktkod. 30460464
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30460464 5 mg
1 options
Produktkod. 30460464 Leverantör LoGiCal Leverantörsnummer LGCFOR0802.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Indoramin Hydrochloride
CAS 38821-52-2
Analyt- eller komponentnamn Indoramin Hydrochloride
Frakt skick Room Temperature
Molekylformel C22 H25 N3 O . Cl H
Certifikat och efterlevnad ISO 9001
Synonym Benzamide, N-[1-[2-(1H-indol-3-yl)ethyl]-4-piperidinyl]-, hydrochloride (1:1), Benzamide, N-[1-[2-(1H-indol-3-yl)ethyl]-4-piperidinyl]-, monohydrochloride (9CI), Baratol, Baratol (pharmaceutical), Doralese, Indoramin hydrochloride, Vidora, Wy 21901 HCl, Wydora, Wypres, Wypresin
InChI formel InChI=1S/C22H25N3O.ClH/c26-22(17-6-2-1-3-7-17)24-19-11-14-25(15-12-19)13-10-18-16-23-21-9-5-4-8-20(18)21;/h1-9,16,19,23H,10-15H2,(H,24,26);1H
Lösningstyp Neat
LEDER Cl.O=C(NC1CCN(CCc2c[nH]c3ccccc23)CC1)c4ccccc4
IUPAC-namn N-[1-[2-(1H-indol-3-yl)ethyl]piperidin-4-yl]benzamide;hydrochloride
Industry Type Clinical Testing
Molekylvikt (g/mol) 383.91
Kvantitet 5 mg
Formel vikt 383.1764 g/mol