Beskrivning
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
- Expert Technical Support
Specifikationer
Specifikationer
| Kemiskt namn eller material | Indoxacarb |
| Förpackning | Glass Bottle |
| CAS | 144171-61-9 |
| Färg | White |
| Smältpunkt | 85°C to 90°C |
| Synonym | Methyl 7-chloro-2,5-dihydro-2-[N-(methoxycarbonyl)-4-(trifluoromethoxy)anilinocarbonyl]indeno[1,2-e][1,3,4]oxadiazine-4a(3H)carboxylate |
| Industry Type | Environmental Analysis |
| Molekylvikt (g/mol) | 527.8 |
| Kvantitet | 25 mg |
| Kvalitet | Analytical Standard |