missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indoxyl-β-D-glucoside, 98%, Thermo Scientific Chemicals
756.93 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 mg
1 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 57.0°C to 58.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (HPLC) |
Quantity | 250 mg |
Solubility Information | Solubility in water: freely soluble. Other solubilities: soluble in methanol, alcohol, acetone, slightly soluble in ether, benzene, chloroform,, ethyl acetate, carbon disulfide |
Formula Weight | 295.29 |
Percent Purity | 98% |
Chemical Name or Material | Indoxyl-β-D-glucoside |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'