Learn More
Cytiva Inlet filter holder kit
Use as filter holder for buffer inlet tubes.   Cytiva Lifescience™ Inlet filter holder kit is an important chromatography accessory.
Brand:  Cytiva 11-0004-07
Additional Details : Weight : 0.02160kg
Product Code. 11340192
  • 1297.00 SEK / Each
Estimated Shipment: 07-05-2024
to see stock.
Add to basket
Description
Description
  • The wetted parts are resistant to organic solvents and salt buffers commonly used in chromatography of biomolecules
  • Except 100 % Ethyl acetate, 100 % Hexane and 100 % Tetrahydrofuran.
  • Support screen Material Support net and filter: Polypropylene (PP)
  • Filter holder: Titanium (Ti) 