Learn More
Eppendorf™ Innova Upright Freezers
Use this freezer for storage and preservation of all types of biological material. New Brunswick Scientific™ Innova upright Freezers combine classical insulation foam with vacuum insulation panels to offer long lasting and reliable ULT technology for safe sample storage.
Brand:  Eppendorf™ U9440-0001
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11379884
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Vacuum insulation panels allow for increased internal capacity with the same footprint as traditionally foamed freezers
  • Inner doors are gasketed as well as insulated creating 3 separate compartments minimizing cold air loss while you access your samples
  • Easy-to-read, eye level, flush mounted LED control panel and alarm status with integrated SMART PLUS™ diagnostics
  • SMART PLUS diagnostic system displays error codes allowing accurate diagnoses by engineers over the phone, saving you time and money
  • Each shelf is adjustable in height for additional storage flexibility
  • Special vacuum vent with plunger ergonomically located at waist level to gain immediate access to your freezer
  • Inner doors can be quickly removed without the need for tools for easier and faster defrosting
  • Full stainless steel interior for easy cleaning during maintenance and defrost cycles; powder-coated steel exterior
  • Special low-temperature outer door seals allow gaskets to remain flexible at –86°C and prevent ice buildup for easy access to samples
  • Password protection of alarm and temperature setpoints to prevent unauthorized setting changes
Specifications
Specifications
-86°C
208 - 230 V
61.5 x 86.5 x 136.5 cm
Vacuum insulation paneling/polyurethane foam
Yes
725 L
Upright
Ultra-low temperature freezers are suitable for storage and preservation of all types of biological materials/samples: Bacteria, cells, Spores, Pollens, Protozoa, Blood components, DNA, Tissue used in research only, Preservation of chemicals or material t
195 cm
U725
102.5 cm
86.7 cm
725 L
60 Hz
315 kg
Yes
725 L
Yes
750 w
Yes
136.5 cm
86.5 cm
Freezer