Learn More
Eppendorf™ Innova Upright Freezers
Use this freezer for storage and preservation of all types of biological material. New Brunswick Scientific™ Innova upright Freezers combine classical insulation foam with vacuum insulation panels to offer long lasting and reliable ULT technology for safe sample storage.
Brand:  Eppendorf™ U9440-0001
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11379884
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance
View alternative products »
Description
Description
  • Vacuum insulation panels allow for increased internal capacity with the same footprint as traditionally foamed freezers
  • Inner doors are gasketed as well as insulated creating 3 separate compartments minimizing cold air loss while you access your samples
  • Easy-to-read, eye level, flush mounted LED control panel and alarm status with integrated SMART PLUS™ diagnostics
  • SMART PLUS diagnostic system displays error codes allowing accurate diagnoses by engineers over the phone, saving you time and money
  • Each shelf is adjustable in height for additional storage flexibility
  • Special vacuum vent with plunger ergonomically located at waist level to gain immediate access to your freezer
  • Inner doors can be quickly removed without the need for tools for easier and faster defrosting
  • Full stainless steel interior for easy cleaning during maintenance and defrost cycles; powder-coated steel exterior
  • Special low-temperature outer door seals allow gaskets to remain flexible at –86°C and prevent ice buildup for easy access to samples
  • Password protection of alarm and temperature setpoints to prevent unauthorized setting changes
Specifications
Specifications
-86°C
208 - 230 V
61.5 x 86.5 x 136.5 cm
Vacuum insulation paneling/polyurethane foam
Yes
725 L
Upright
Ultra-low temperature freezers are suitable for storage and preservation of all types of biological materials/samples: Bacteria, cells, Spores, Pollens, Protozoa, Blood components, DNA, Tissue used in research only, Preservation of chemicals or material t
195 cm
U725
102.5 cm
86.7 cm
725 L
60 Hz
315 kg
Yes
725 L
Yes
750 w
Yes
136.5 cm
86.5 cm
Freezer
Videos
Documents
View accessory products