Learn More
Eppendorf™ Innova Upright Freezers
Use this freezer for storage and preservation of all types of biological material. New Brunswick Scientific™ Innova upright Freezers combine classical insulation foam with vacuum insulation panels to offer long lasting and reliable ULT technology for safe sample storage.
73500.00 SEK - 118805.00 SEK
Specifications
For Use With (Equipment) Ultra-low temperature freezers are suitable for storage and preservation of all types of biological materials/samples: Bacteria, cells, Spores, Pollens, Protozoa, Blood components, DNA, Tissue used in research only, Preservation of chemicals or material t
Door Shelves Yes
Insulation Vacuum insulation paneling/polyurethane foam
Shelves Yes
Type Freezer
View More Specs
Products 3
Product Code Brand Dimensions (D x W x H) Interior Price Quantity & Availability  
Product Code Brand Dimensions (D x W x H) Interior Price Quantity & Availability  
11321082 View accessory products View Documents Eppendorf™
U9420-0001
33 x 48 x 64 cm 73500.00 SEK
Each View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 13-11-2019
Add to basket
11369884 View accessory products View Documents Eppendorf™
U9430-0001
61.5 x 64 x 136.5 cm 113800.00 SEK
Each View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 11-11-2019
Add to basket
11379884 View accessory products View Documents Eppendorf™
U9440-0001
61.5 x 86.5 x 136.5 cm This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance
N/A View alternative products
Save Up to 
»
Description
Description
  • Vacuum insulation panels allow for increased internal capacity with the same footprint as traditionally foamed freezers
  • Inner doors are gasketed as well as insulated creating 3 separate compartments minimizing cold air loss while you access your samples
  • Easy-to-read, eye level, flush mounted LED control panel and alarm status with integrated SMART PLUS™ diagnostics
  • SMART PLUS diagnostic system displays error codes allowing accurate diagnoses by engineers over the phone, saving you time and money
  • Each shelf is adjustable in height for additional storage flexibility
  • Special vacuum vent with plunger ergonomically located at waist level to gain immediate access to your freezer
  • Inner doors can be quickly removed without the need for tools for easier and faster defrosting
  • Full stainless steel interior for easy cleaning during maintenance and defrost cycles; powder-coated steel exterior
  • Special low-temperature outer door seals allow gaskets to remain flexible at –86°C and prevent ice buildup for easy access to samples
  • Password protection of alarm and temperature setpoints to prevent unauthorized setting changes
Specifications
Specifications
Ultra-low temperature freezers are suitable for storage and preservation of all types of biological materials/samples: Bacteria, cells, Spores, Pollens, Protozoa, Blood components, DNA, Tissue used in research only, Preservation of chemicals or material t
Vacuum insulation paneling/polyurethane foam
Freezer
-86°C