missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INO80 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18810-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
DNA helicase and probable main scaffold component of the chromatin remodeling INO80 complex which is involved in transcriptional regulation, DNA replication and probably DNA repair; according to PubMed:20687897 the contribution to DNA double-strand break repair appears to be largely indirect through transcriptional regulation. Recruited by YY1 to YY1-activated genes, where it acts as an essential coactivator. Binds DNA. In vitro, has double-stranded DNA-dependent ATPase activity. Involved in UV-damage excision repair, DNA replication and chromosome segregation during normal cell division cycle.Specifications
INO80 | |
Polyclonal | |
Unconjugated | |
INO80 | |
hINO80, INO80, INO80 homolog (S. cerevisiae), INO80A, INOC1, KIAA1259 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
296084, 54617, 68142 | |
-20°C | |
Liquid |
Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6ZPV2, Q9ULG1 | |
INO80 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |