missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INO80C Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24793-1-AP-20UL
Additional Details : Weight : 0.00010kg
Specifications
INO80C | |
Polyclonal | |
Unconjugated | |
INO80C | |
C18orf37, hIes6, IES6, INO80 complex subunit C, INO80C | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
125476 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.56 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6PI98 | |
INO80C | |
INO80C Fusion Protein Ag20468 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |