Läs mer
Inosine-5'-monophosphate Sodium Salt Hydrate, TRC
Produktkod. 30008004
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008004 100 mg
30006355 50 mg
30010991 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30008004 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI665010100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 20813-76-7
Molekylformel C10 H11 N4 O8 P . 2[Na] . 8[H2 O]
Molekylvikt (g/mol) 536.292
IUPAC-namn disodium;[(2R,3S,4R,5R)-3,4-dihydroxy-5-(6-oxo-1H-purin-9-yl)oxolan-2-yl]methyl phosphate;octahydrate
LEDER O.O.O.O.O.O.O.O.[Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP(=O)([O-])[O-])n2cnc3C(=O)NC=Nc23
Specifikationer
Kemiskt namn eller material Inosine-5'-monophosphate Sodium Salt Hydrate
Kvantitet 100 mg
InChI formel InChI=1S/C10H13N4O8P.2Na.8H2O/c15-6-4(1-21-23(18,19)20)22-10(7(6)16)14-3-13-5-8(14)11-2-12-9(5)17;;;;;;;;;;/h2-4,6-7,10,15-16H,1H2,(H,11,12,17)(H2,18,19,20);;;8*1H2/q;2*+1;;;;;;;;/p-2/t4-,6-,7-,10-;;;;;;;;;;/m1........../s1
Formel vikt 536.096