missing translation for 'onlineSavingsMsg'
Learn More

INPP5B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15141-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16824194

  • 3313.85 SEK / 150µL
Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Cellular calcium signaling is controlled by the production of inositol phosphates (IPs) by phospholipase C in response to extracellular signals. The IP signaling molecules are inactivated by a family of inositol polyphosphate-5-phosphatases (5-phosphatases). INPP5B encodes the type II 5-phosphatase. The protein is localized to the cytosol and mitochondria, and associates with membranes through an isoprenyl modification near the C-terminus. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
Specifications

Specifications

INPP5B
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P32019, Q8K337
5PTase, INPP5B, Phosphoinositide 5 phosphatase
Rabbit
IgG
150 μL
-20°C
Primary
16330, 362590, 3633
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
INPP5B
Liquid
Inpp5b
INPP5B Fusion Protein Ag7315
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers