Learn More
Ins1 Mouse anti-Rat, Unconjugated, Clone: 3A11C1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67284-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16839825
  • 3313.85 SEK / 150µL
Estimated Shipment: 05-10-2022
to see stock.
Description
Description
The Ins1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of rat Ins1. This antibody recognizes rat, mouse antigen. The Ins1 antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
Specifications
Ins1
Monoclonal
1000 μg/mL
Mouse
Primary
Mouse, Rat
ELISA, Immunofluorescence, Immunohistochemistry
3A11C1
Unconjugated
150 μL
24505
IgG2a