Läs mer
Brady™ Inspection Timer
Klicka för att se tillgängliga alternativ
Tidsintervall:
30 dagar
365 dagar
7 dagar.
Förpackningsstorlek:
Paket med 10
Beskrivning
- Clearly highlights when the next planned maintenance intervention is due
- Versions are available that count down 7 days, 30 days, 180 days and 365 days
- Each version is equipped with a dark green, light green, yellow and flashing red LED light that indicate a recently inspected up to an uncertain equipment status
Specifikationer
Specifikationer
| Tidsintervall | 30 dagar |
| Visa | LED |
| Batterityp | (3) AAA Alkaline |
| Vikt (metrisk) | 1030 g |