missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ INSR Recombinant Human Protein
INSR is a receptor tyrosine kinase.
Brand: Invitrogen™ PR7080A
Additional Details : Weight : 0.01000kg
Description
Binding of insulin to the extracellular domain of INSR stimulates glucose uptake.
Specifications
NP_000199 | |
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
INSR | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
71.5kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Cytoplasmic | |
Insulin & IGF Receptor Kinase Family | |
Recombinant | |
Baculovirus |