missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INSRR (IRR) Recombinant Human Protein
INSRR (IRR) Recombinant Human Protein
Brand: Invitrogen PV3808
Additional Details : Weight : 0.05250kg
Description
INSRR, insulin receptor-related receptor, is a member of the insulin receptor (IR) family. INSRR does not bind insulin-like molecules, but is similar to INSR in tyrosine kinase activity.Specifications
NP_055030 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
67.1kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
INSRR (IRR), IRR | |
INSRR (IRR) was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL AXLtide peptide substrate (KKSRGDYMTMQIG) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
Amino acids 944-1297 | |
990nmoles of phosphate transferred to AXLtide peptide substrate (KKSRGDYMTMQIG) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 1.67μg/mL | |
GST Tag | |
Kinase and inhibitors | |
90% by SDS-PAGE |
0.39mg/mL | |
3645 | |
INSRR (IRR) Recombinant Human Protein | |
–80°C | |
1 Each | |
INSRR (IRR) Recombinant Human Protein | |
Insulin & IGF receptor | |
INSRR (IRR) Recombinant Human Protein | |
Human | |
Baculovirus | |
Receptor tyrosine kinases |
For Research Use Only. Not for use in diagnostic procedures.